(1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene)copper(III) chloride 95%

Supplier: Ambeed
Warning

A1501169-1G A1501169-250MG A1501169-100MG
76666-922EA 240.83 USD
76666-922 76666-924 77624-662
(1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene)copper(III) chloride 95%
(1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene)copper(III) chloride

Precaución: for Research Use Only

Formula: C₂₇H₃₈ClCuN₂
MW: 489.6030 g/mol
CAS Number: 915395-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR