(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) 97%

Supplier: Ambeed
Warning

A1501030-100MG A1501030-1G A1501030-250MG
76666-904EA 222.79 USD
76666-904 76666-906 76666-908
(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) 97%
(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₅₅H₅₈N₂O₂
MW: 779.0612 g/mol
CAS Number: 2757085-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR