(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) 97%

Supplier: Ambeed
Warning

A1501029-100MG A1501029-1G A1501029-250MG
76666-898EA 222.79 USD
76666-898 76666-900 76666-902
(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) 97%
(3aS,3a'S,8aR,8a'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₅₅H₅₈N₂O₂
MW: 779.0612 g/mol
CAS Number: 2757085-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR