1-(tert-Butoxy)-4-ethylbenzene 98%

Supplier: Ambeed
Warning

A1500278-5G A1500278-1G A1500278-250MG A1500278-25G
76666-860EA 371 USD
76666-860 77624-652 77875-707 77875-708
1-(tert-Butoxy)-4-ethylbenzene 98%
1-(tert-Butoxy)-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
CAS Number: 215394-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR