(S)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride 96+%

Supplier: Ambeed
Warning

A1499355-100MG A1499355-1G A1499355-250MG
76666-854EA 68.96 USD
76666-854 76666-856 76666-858
(S)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride 96+%
(S)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₂Cl₃N₃
MW: 278.6501 g/mol
CAS Number: 2512230-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR