(R)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride ≥96%

Supplier: Ambeed
Warning

A1499351-100MG A1499351-1G A1499351-250MG
76666-848EA 66.84 USD
76666-848 76666-850 76666-852
(R)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride ≥96%
(R)-1-Methyl-4-(pyrrolidin-3-yl)piperazine trihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₂Cl₃N₃
MW: 278.6501 g/mol
MDL Number: MFCD28023885
CAS Number: 917505-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR