Tri-tert-butyl 2,2',2''-(10-(2-((4-aminobutyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 95%
Supplier: Ambeed
|
Warning
|
A1499166-100MG
A1499166-1G
A1499166-250MG
A1499166-50MG
76666-832EA
251
USD
76666-832
76666-834
76666-836
76666-838
Tri-tert-butyl 2,2',2''-(10-(2-((4-aminobutyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 95%
Tri-tert-butyl 2,2',2''-(10-(2-((4-aminobutyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate
Precaución: for Research Use Only
Formula:
C₃₂H₆₂N₆O₇ |
CAS Number:
1402393-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...