5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%

Supplier: Ambeed
Warning

A1499119-1G A1499119-250MG A1499119-5G
76666-816EA 13 USD
76666-816 76666-818 77624-618
5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%
5-Bromo-3-fluoro-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR