Bis(hexafluoroacetylacetonato)cobalt(II) hydrate 98% + (contains < 12% H₂O)

Supplier: Ambeed
Warning

Synonyms: Hexafluoroacetylacetono Cobalt(II) Salt, Cobalt(II) Hexafluoroacetylacetonate

A1499047-1G A1499047-250MG A1499047-5G
76666-810EA 24 USD
76666-810 76666-812 76666-814
Bis(hexafluoroacetylacetonato)cobalt(II) hydrate 98% + (contains < 12% H₂O)
Bis(hexafluoroacetylacetonato)cobalt(II) hydrate

Precaución: for Research Use Only

Formula: C₁₀H₂CoF₁₂O₄
MW: 473.04 g/mol
MDL Number: MFCD00042510
CAS Number: 19648-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR