1-Tetradecyl-3-methylimidazolium chloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-3-tetradecyl-1H-imidazol-3-ium chloride

A1497710-25G A1497710-5G
76666-786EA 22.28 USD
76666-786 76666-788
1-Tetradecyl-3-methylimidazolium chloride 95%
1-Tetradecyl-3-methylimidazolium chloride

Precaución: for Research Use Only

Formula: C₁₈H₃₅ClN₂
MW: 314.9369 g/mol
MDL Number: MFCD03427607
CAS Number: 171058-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR