2-Fluoro-8-nitro-3,5,6,7-tetrahydro-s-indacen-1(2H)-one 97%
Supplier: Ambeed
|
Warning
|
A1497645-100MG
A1497645-250MG
76666-774EA
53.05
USD
76666-774
76666-776
2-Fluoro-8-nitro-3,5,6,7-tetrahydro-s-indacen-1(2H)-one 97%
2-Fluoro-8-nitro-3,5,6,7-tetrahydro-s-indacen-1(2H)-one
Precaución: for Research Use Only
Formula:
C₁₂H₁₀FNO₃ MW: 235.2111 g/mol |
CAS Number:
2676867-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...