6-Fluoro-4-nitro-2,3-dihydro-1H-inden-5-ol 97%

Supplier: Ambeed
Warning

A1497625-100MG A1497625-250MG
76666-756EA 27 USD
76666-756 76666-758
6-Fluoro-4-nitro-2,3-dihydro-1H-inden-5-ol 97%
6-Fluoro-4-nitro-2,3-dihydro-1H-inden-5-ol

Precaución: for Research Use Only

Formula: C₉H₈FNO₃
MW: 197.1631 g/mol
CAS Number: 2676865-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR