(4-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-2-yl)methanol 97%

Supplier: Ambeed
Warning

A1497617-100MG A1497617-250MG
76666-750EA 98.67 USD
76666-750 76666-752
(4-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-2-yl)methanol 97%
(4-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.26 g/mol
CAS Number: 2676865-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR