(8-Amino-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol 97%

Supplier: Ambeed
Warning

A1497606-100MG A1497606-250MG
76666-742EA 51 USD
76666-742 76666-744
(8-Amino-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol 97%
(8-Amino-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO
CAS Number: 2676864-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR