7-Amino-6-cyclopropyl-2,3-dihydro-1H-indene-4-carbonitrile 97%

Supplier: Ambeed
Warning

A1497588-100MG A1497588-1G A1497588-250MG
76666-728EA 53.05 USD
76666-728 76666-730 76666-732
7-Amino-6-cyclopropyl-2,3-dihydro-1H-indene-4-carbonitrile 97%
7-Amino-6-cyclopropyl-2,3-dihydro-1H-indene-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂
MW: 198.26 g/mol
CAS Number: 2676863-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR