5-Bromo-2,3-dihydro-1H-inden-4-ol 97%

Supplier: Ambeed
Warning

A1497581-100MG A1497581-250MG
76666-720EA 26.53 USD
76666-720 76666-722
5-Bromo-2,3-dihydro-1H-inden-4-ol 97%
5-Bromo-2,3-dihydro-1H-inden-4-ol

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.0712 g/mol
MDL Number: MFCD23133290
CAS Number: 575504-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR