8-Nitro-1-(trifluoromethyl)-1,2,3,5,6,7-hexahydro-s-indacen-1-ol 97%
Supplier: Ambeed
|
Warning
|
A1497571-100MG
A1497571-250MG
76666-706EA
120.95
USD
76666-706
76666-708
8-Nitro-1-(trifluoromethyl)-1,2,3,5,6,7-hexahydro-s-indacen-1-ol 97%
8-Nitro-1-(trifluoromethyl)-1,2,3,5,6,7-hexahydro-s-indacen-1-ol
Precaución: for Research Use Only
Formula:
C₁₃H₁₂F₃NO₃ MW: 287.2345 g/mol |
CAS Number:
2676863-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...