3-Isopropyl-1-methoxy-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A1497556-100MG
A1497556-250MG
76666-690EA
89.12
USD
76666-690
76666-692
3-Isopropyl-1-methoxy-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%
3-Isopropyl-1-methoxy-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂O MW: 206.28 g/mol |
CAS Number:
2676862-51-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...