3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A1497554-100MG
A1497554-250MG
76666-686EA
51.99
USD
76666-686
76666-688
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine
Precaución: for Research Use Only
Formula:
C₁₁H₁₆N₂ MW: 176.25 g/mol |
CAS Number:
2409124-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...