2-Hydroxypropyl methacrylate (mixture of isomers) 97% + stabilised

Supplier: Ambeed
Warning

Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1497522-100G A1497522-25G A1497522-500G
76666-678EA 7 USD
76666-678 76666-680 76666-682
2-Hydroxypropyl methacrylate (mixture of isomers) 97% + stabilised
2-Hydroxypropyl methacrylate (mixture of isomers)

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00004536
CAS Number: 27813-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR