rel-(3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol 98+%
Supplier: Ambeed
|
Warning
|
A1497324-1G
A1497324-25G
76666-674EA
119.89
USD
76666-674
76666-676
rel-(3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol 98+%
rel-(3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
Precaución: for Research Use Only
Formula:
C₆H₁₂O₆ MW: 180.15 g/mol |
CAS Number:
54-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...