Ethyl 5-amino-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A1497271-100G A1497271-25G
76666-670EA 163.38 USD
76666-670 77624-606
Ethyl 5-amino-2-methylbenzoate 98%
Ethyl 5-amino-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
MDL Number: MFCD11226196
CAS Number: 7335-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR