2-((3-Cyano-6-oxo-4-(p-tolyl)-1,4,5,6-tetrahydropyridin-2-yl)thio)-N-(thiazol-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A1497217-25MG A1497217-100MG A1497217-250MG A1497217-50MG
76666-666EA 112 USD
76666-666 77875-664 77875-665 77875-666
2-((3-Cyano-6-oxo-4-(p-tolyl)-1,4,5,6-tetrahydropyridin-2-yl)thio)-N-(thiazol-2-yl)acetamide 98%
2-((3-Cyano-6-oxo-4-(p-tolyl)-1,4,5,6-tetrahydropyridin-2-yl)thio)-N-(thiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₄O₂S₂
MDL Number: MFCD02241236
CAS Number: 375835-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR