5,10,15,20-Tetrakis(3-bromophenyl)porphyrin 95%

Supplier: Ambeed
Warning

A1496611-100MG A1496611-1G A1496611-250MG
76666-652EA 85.94 USD
76666-652 76666-654 76666-656
5,10,15,20-Tetrakis(3-bromophenyl)porphyrin 95%
5,10,15,20-Tetrakis(3-bromophenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₄H₂₆Br₄N₄
MW: 930.3201 g/mol
CAS Number: 68772-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR