tert-Butyl (octahydrocyclopenta[c]pyrrol-5-yl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A1496471-100MG A1496471-250MG
76666-648EA 105 USD
76666-648 76666-650
tert-Butyl (octahydrocyclopenta[c]pyrrol-5-yl)carbamate hydrochloride 95%
tert-Butyl (octahydrocyclopenta[c]pyrrol-5-yl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₃ClN₂O₂
CAS Number: 2757731-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR