1,3-Dioxoisoindolin-2-yl dodecanoate 95%
Supplier: Ambeed
|
Warning
|
A1496223-1G
A1496223-250MG
A1496223-5G
76666-618EA
73.21
USD
76666-618
76666-620
76666-622
1,3-Dioxoisoindolin-2-yl dodecanoate 95%
1,3-Dioxoisoindolin-2-yl dodecanoate
Precaución: for Research Use Only
Formula:
C₂₀H₂₇NO₄ MW: 345.43 g/mol |
CAS Number:
1947316-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...