1,3-Dioxoisoindolin-2-yl benzoate 97%

Supplier: Ambeed
Warning

A1496221-1G A1496221-25G A1496221-5G
76666-612EA 28.65 USD
76666-612 76666-614 76666-616
1,3-Dioxoisoindolin-2-yl benzoate 97%
1,3-Dioxoisoindolin-2-yl benzoate

Precaución: for Research Use Only

Formula: C₁₅H₉NO₄
MW: 267.2363 g/mol
CAS Number: 58585-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR