1,3-Dioxoisoindolin-2-yl 2-propylpentanoate 97%

Supplier: Ambeed
Warning

A1496218-1G A1496218-250MG A1496218-5G
76666-602EA 47 USD
76666-602 76666-604 76666-606
1,3-Dioxoisoindolin-2-yl 2-propylpentanoate 97%
1,3-Dioxoisoindolin-2-yl 2-propylpentanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₄
MW: 289.3264 g/mol
CAS Number: 2387417-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR