1,3-Dioxoisoindolin-2-yl 2,2-dimethylbutanoate 97%

Supplier: Ambeed
Warning

A1496216-1G A1496216-5G
76666-592EA 48.81 USD
76666-592 76666-594
1,3-Dioxoisoindolin-2-yl 2,2-dimethylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 2,2-dimethylbutanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
MW: 261.27 g/mol
CAS Number: 1928767-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR