1,3-Dioxoisoindolin-2-yl adamantane-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A1496215-1G
A1496215-25G
A1496215-5G
76666-586EA
45
USD
76666-586
76666-588
76666-590
1,3-Dioxoisoindolin-2-yl adamantane-1-carboxylate 97%
1,3-Dioxoisoindolin-2-yl adamantane-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₉H₁₉NO₄ |
CAS Number:
118334-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...