1,3-Dioxoisoindolin-2-yl 2-methyl-2-phenylpropanoate 97%

Supplier: Ambeed
Warning

A1496214-1G A1496214-250MG A1496214-5G
76666-580EA 86 USD
76666-580 76666-582 76666-584
1,3-Dioxoisoindolin-2-yl 2-methyl-2-phenylpropanoate 97%
1,3-Dioxoisoindolin-2-yl 2-methyl-2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₄
CAS Number: 2241846-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR