1,3-Dioxoisoindolin-2-yl 2-phenoxyacetate 97%

Supplier: Ambeed
Warning

A1496212-1G A1496212-250MG A1496212-5G
76666-574EA 68 USD
76666-574 76666-576 76666-578
1,3-Dioxoisoindolin-2-yl 2-phenoxyacetate 97%
1,3-Dioxoisoindolin-2-yl 2-phenoxyacetate

Precaución: for Research Use Only

Formula: C₁₆H₁₁NO₅
CAS Number: 109524-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR