1,3-Dioxoisoindolin-2-yl tetrahydro-2H-pyran-4-carboxylate 97%

Supplier: Ambeed
Warning

A1496210-1G A1496210-250MG A1496210-5G
76666-568EA 83.82 USD
76666-568 76666-570 76666-572
1,3-Dioxoisoindolin-2-yl tetrahydro-2H-pyran-4-carboxylate 97%
1,3-Dioxoisoindolin-2-yl tetrahydro-2H-pyran-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₅
MW: 275.2567 g/mol
CAS Number: 1872262-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR