1,3-Dioxoisoindolin-2-yl 2-(4-fluorophenyl)acetate 97%

Supplier: Ambeed
Warning

A1496206-1G A1496206-250MG
76666-548EA 35 USD
76666-548 76666-550
1,3-Dioxoisoindolin-2-yl 2-(4-fluorophenyl)acetate 97%
1,3-Dioxoisoindolin-2-yl 2-(4-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₀FNO₄
CAS Number: 1960404-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR