1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
A1496205-1G
A1496205-250MG
A1496205-5G
76666-542EA
73
USD
76666-542
76666-544
76666-546
1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate 97%
1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₉H₂₂N₂O₆ |
MDL Number:
MFCD32862308 CAS Number: 1872262-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...