1,3-Dioxoisoindolin-2-yl cyclopentanecarboxylate 97%

Supplier: Ambeed
Warning

A1496204-1G A1496204-250MG A1496204-5G
76666-536EA 83.82 USD
76666-536 76666-538 76666-540
1,3-Dioxoisoindolin-2-yl cyclopentanecarboxylate 97%
1,3-Dioxoisoindolin-2-yl cyclopentanecarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₄
MW: 259.2573 g/mol
CAS Number: 946513-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR