1,3-Dioxoisoindolin-2-yl cyclobutanecarboxylate 97%

Supplier: Ambeed
Warning

A1496203-1G A1496203-5G A1496203-250MG
76666-532EA 73 USD
76666-532 76666-534 77875-640
1,3-Dioxoisoindolin-2-yl cyclobutanecarboxylate 97%
1,3-Dioxoisoindolin-2-yl cyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₄
CAS Number: 1872262-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR