1,3-Dioxoisoindolin-2-yl 3-phenylpropanoate 97%

Supplier: Ambeed
Warning

A1496201-1G A1496201-250MG A1496201-5G
76666-522EA 65 USD
76666-522 76666-524 76666-526
1,3-Dioxoisoindolin-2-yl 3-phenylpropanoate 97%
1,3-Dioxoisoindolin-2-yl 3-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₇H₁₃NO₄
CAS Number: 84379-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR