1,3-Dioxoisoindolin-2-yl 4-phenylbutanoate 97%

Supplier: Ambeed
Warning

A1496200-1G A1496200-250MG A1496200-5G
76666-516EA 76 USD
76666-516 76666-518 76666-520
1,3-Dioxoisoindolin-2-yl 4-phenylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 4-phenylbutanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₄
CAS Number: 1841512-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR