1,3-Dioxoisoindolin-2-yl 2-methylhexanoate 97%
Supplier: Ambeed
|
Warning
|
A1496199-1G
A1496199-250MG
A1496199-5G
76666-510EA
47
USD
76666-510
76666-512
76666-514
1,3-Dioxoisoindolin-2-yl 2-methylhexanoate 97%
1,3-Dioxoisoindolin-2-yl 2-methylhexanoate
Precaución: for Research Use Only
Formula:
C₁₅H₁₇NO₄ |
CAS Number:
1967011-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...