1,3-Dioxoisoindolin-2-yl 2-methylpentanoate 97%

Supplier: Ambeed
Warning

A1496197-1G A1496197-250MG A1496197-5G
76666-498EA 36 USD
76666-498 76666-500 76666-502
1,3-Dioxoisoindolin-2-yl 2-methylpentanoate 97%
1,3-Dioxoisoindolin-2-yl 2-methylpentanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
CAS Number: 2170681-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR