1,3-Dioxoisoindolin-2-yl 2-ethylhexanoate 97%

Supplier: Ambeed
Warning

A1496196-1G A1496196-250MG A1496196-5G
76666-492EA 35 USD
76666-492 76666-494 76666-496
1,3-Dioxoisoindolin-2-yl 2-ethylhexanoate 97%
1,3-Dioxoisoindolin-2-yl 2-ethylhexanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₄
CAS Number: 1872262-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR