1,3-Dioxoisoindolin-2-yl 2-methylbutanoate 97%

Supplier: Ambeed
Warning

A1496195-1G A1496195-250MG A1496195-5G
76666-486EA 35 USD
76666-486 76666-488 76666-490
1,3-Dioxoisoindolin-2-yl 2-methylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 2-methylbutanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
CAS Number: 2171537-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR