1,3-Dioxoisoindolin-2-yl 3,3-dimethylbutanoate 97%

Supplier: Ambeed
Warning

A1496193-1G A1496193-250MG A1496193-5G
76666-474EA 58 USD
76666-474 76666-476 76666-478
1,3-Dioxoisoindolin-2-yl 3,3-dimethylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 3,3-dimethylbutanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
CAS Number: 1894176-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR