1,3-Dioxoisoindolin-2-yl 3-methylbutanoate 97%

Supplier: Ambeed
Warning

A1496192-1G A1496192-250MG A1496192-5G
76666-468EA 34 USD
76666-468 76666-470 76666-472
1,3-Dioxoisoindolin-2-yl 3-methylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 3-methylbutanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
MW: 247.2466 g/mol
CAS Number: 2125731-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR