1,3-Dioxoisoindolin-2-yl butyrate 97%

Supplier: Ambeed
Warning

A1496191-1G A1496191-25G A1496191-5G
76666-462EA 55 USD
76666-462 76666-464 76666-466
1,3-Dioxoisoindolin-2-yl butyrate 97%
1,3-Dioxoisoindolin-2-yl butyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₄
MW: 233.2200 g/mol
CAS Number: 1538551-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR