1,12-Dodecanediol 98%

Supplier: Ambeed
Warning

A1495981-100G A1495981-25G A1495981-500G
76666-456EA 44 USD
76666-456 76666-458 76666-460
1,12-Dodecanediol 98%
1,12-Dodecanediol

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₂
MDL Number: MFCD00004755
CAS Number: 5675-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR