tert-Butyl 4-bromobutanoate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-bromobutyrate

A1495831-100G A1495831-25G A1495831-500G A1495831-5G
76666-446EA 139 USD
76666-446 76666-448 76666-450 76666-452
tert-Butyl 4-bromobutanoate 95%
tert-Butyl 4-bromobutanoate

Precaución: for Research Use Only

Formula: C₈H₁₅BrO₂
MW: 223.107 g/mol
MDL Number: MFCD02179416
CAS Number: 110661-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR