5,7-Di-tert-butyl-3-phenylbenzo[d]oxazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1495767-1G A1495767-250MG A1495767-5G A1495767-25G A1495767-100MG
76666-440EA 45 USD
76666-440 76666-442 76666-444 77549-864 77597-700
5,7-Di-tert-butyl-3-phenylbenzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-phenylbenzo[d]oxazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₆BF₄NO
MDL Number: MFCD34566941
CAS Number: 1207294-92-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR