[5,5'-Bis-trifluoromethyl]bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495459-250MG A1495459-50MG A1495459-100MG A1495459-1G
76666-430EA 432 USD
76666-430 76666-432 77875-630 77875-631
[5,5'-Bis-trifluoromethyl]bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%
[5,5'-Bis-trifluoromethyl]bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₁₆F₁₈IrN₄P
CAS Number: 2757085-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR